missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Piperonyl isobutyrate, 99%

Product Code. 11429237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429237 100 g 100g
11419237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429237 Supplier Alfa Aesar™ Supplier No. B21321.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5461-08-5
MDL Number MFCD00005826

Specifications

Density 1.154
Boiling Point 91°C to 92°C (0.005 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C12H14O4
Refractive Index 1.512
Quantity 100 g
Synonym piperonyl isobutyrate, unii-y8hbr1acya, y8hbr1acya, piperonyl 2-methylpropanoate, heliotropyl 2-methylpropanoate, isobutyric acid piperonyl ester, propanoic acid, 2-methyl-, 1,3-benzodioxol-5-ylmethyl ester, 1,3-benzodioxol-5-ylmethyl isobutyrate, 3,4-methylenedioxybenzyl 2-methylpropanoate, isobutyric acid 3,4-methylenedioxybenzyl ester
InChI Key RQULTIASPCVEFO-UHFFFAOYSA-N
SMILES CC(C)C(=O)OCC1=CC2=C(C=C1)OCO2
IUPAC Name 1,3-benzodioxol-5-ylmethyl 2-methylpropanoate
Molecular Weight (g/mol) 222.24
PubChem CID 62580
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Piperonyl isobutyrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.