missing translation for 'onlineSavingsMsg'
Learn More

Piperonylic acid, 98+%

Product Code. 11458056
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458056 100 g 100g
11448056 25 g 25g
11468056 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458056 Supplier Thermo Scientific Alfa Aesar Supplier No. A14105.22
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 94-53-1 | C8H6O4 | 166.13 g/mol

Piperonylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-53-1
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00005830
InChI Key VDVJGIYXDVPQLP-UHFFFAOYSA-N
Synonym piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid
PubChem CID 7196
IUPAC Name 1,3-benzodioxole-5-carboxylic acid
SMILES OC(=O)C1=CC=C2OCOC2=C1

Specifications

Melting Point 230°C to 235°C
Quantity 100 g
Beilstein 150206
Merck Index 14,7477
Solubility Information Slightly soluble in water.
Formula Weight 166.13
Percent Purity ≥98%
Chemical Name or Material Piperonylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.