missing translation for 'onlineSavingsMsg'
Learn More

Piperonylonitrile, 97%, Thermo Scientific™

Product Code. 10103110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103110 5 g Glass bottle 5g
10735991 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103110 Supplier Thermo Scientific Acros Supplier No. 151880050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4421-09-4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00005820
InChI Key PKRWWZCDLJSJIF-UHFFFAOYSA-N
Synonym piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358
PubChem CID 78136
IUPAC Name 1,3-benzodioxole-5-carbonitrile
SMILES C1OC2=C(O1)C=C(C=C2)C#N

Specifications

Melting Point 90.0°C to 95.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H5NO2
Quantity 5 g
Beilstein 19, 270
Formula Weight 147.13
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Piperonylonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Piperonylonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.