Learn More
Sartorius Pipet Charging Stand and Carousel
Product Code. 10191113 Shop All Sartorius Products
Change view
Click to view available options
Number of Channels:
1
4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Number of Channels Unit Size
10191113 4 Each
10670744 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10191113 Supplier Sartorius Supplier No. 730991
  • ONLINE EXCLUSIVE PRICE
    1024.95 €
    655.00 € / Pack of 1
    Save 369.95 €
    36% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Includes
Universal AC-adaptor
Saves bench space. Sartorius Pipet Charging Stand and Carousel holds and stores pipets in an upright position to avoid contamination. For use with PICUS™ and eLINE™ mechanical and electronic pipets.
For storing and charging Picus and eLINE mechanical and electronic pipets when not in use. Pipets should be stored in an upright position in order to avoid contamination from desks and benches.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipette Tips
Material Polypropylene
Number of Channels 4
Type Pipette Holder
For Use With (Equipment) Picus and eLINE Electronic Pipettes
Includes Universal AC adapter