missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pipette Tips, 2 to 200 μL
GREENER_CHOICE

Product Code. 15174897 Shop All Brand Products
Change view
Click to view available options
Packaging:
Bagged (1000 Pk.)
Tip-Stack (10 x 96 Tips)
TipBox
TipRack
Unit Size:
Pack of 1000
Pack of 480
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
15174897 TipRack Pack of 960
15154887 Bagged (1000 Pk.) Pack of 1000
15124927 Tip-Stack (10 x 96 Tips) Pack of 960
15104917 TipBox Pack of 480
15134897 Bagged (1000 Pk.) Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15174897 Supplier BRAND™ Supplier No. 732108
Stock Up and Save on BRAND Lab Essentials
Use promo code 27939
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed for high-quality micro-pipettes and suitable for various applications. BRAND™ 2 to 200 μL pipette tips ensure precise and efficient pipetting. Tips are cleanroom-produced, free from contaminants, and environmentally friendly.

  • Engineered to fit the Transferpette™ line of micropipettes
  • Compatibility tested with other micropipettors from various manufacturers
  • Manufactured and packaged under certified cleanroom conditions according to ISO 14644-1, Class 7 and 8
  • Made from high-quality, pure raw materials, free of DiHEMDA and oleamide
  • No use of mold release agents such as stearates and erucamide in the production process
  • Palletized BIO-CERT™ quality tips are guaranteed to be free from DNA, DNase, RNase, ATP, and endotoxins
  • Thin-walled for high translucency, easy ejection, and low waste
  • Graduations at 20 and 100 μL for quick volume checks
  • Available either in bulk packaging or convenient, ecological rack-and-stack formats
  • Bulk tips are yellow and placed in clear, resealable bags
  • Racked tips are colorless and placed into yellow-colored tip trays
  • TipBoxes are made of 100% PP and are fully recyclable
  • The TipRack refilling system reduces waste by more than 20% compared to the TipBox
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 2 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Refill
Tip Style Universal
Packaging TipRack
Product Type Pipette Tips
Color Code Yellow
Disposable Limited-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 20, 100 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.