Learn More
ratiolab™ Pipet Tips
Product Code. 15110386
Click to view available options
Color:
Blue
Clear
Yellow
Sterility:
Non-sterile
Sterile
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
1 to 250 μL
100 to 1000 μL
100 to 1200 μL
500 to 2500 μL
Packaging:
Bag
Multirack
Unit Size:
Pack of 1000
Pack of 250
Pack of 600
Pack of 960
Product Code. 15110386
Brand: ratiolab™ 2553172
Only null left
Add to basket
Add to basket
Pipet tips for all commercially available manual or electronic, single or multichannel pipets.
  • Guaranteed dust-free due to special ionization treatment
  • High transparency due to high quality surfaces
  • Minimal residual wetting due to the controlled quality of raw materials
  • High volume accuracy due to precise tip orifice
  • Together with specially developed Multirack, these make pipetting much simpler and safer in the most sensitive applications
Specifications
Volume (Metric) 0.5 to 20 μL
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Packaging Multirack
Quantity 10 x 96 Tips/Pk.
Material Polypropylene