missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ Comfort Handle, White

Product Code. 15392589 Shop All Gilson Products
Change view
Click to view available options
Quantity:
3 Handles
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15392589 3 Handles Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15392589 Supplier Gilson™ Supplier No. F161488
  • ONLINE EXCLUSIVE PRICE
    55.35 €
    53.87 € / Pack of 3
    Save 1.48 €
    2% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ergonomically designed PIPETMAN™ Comfort Handle attaches to any PIPETMAN™ Classic, Neo, or G pipette to provide improved comfort and reduce pipetting fatigue. The handles are easily personalized using the PIPETMAN™ Comfort Handle Labels.

  • Large, comfortable finger hook and ergonomically designed handle offers a better grip to support daily pipetting activity and reduces the risk of repetitive strain injuries (RSI)
  • Mark with the included pre-cut adhesive labels, designed with the online Label Maker Tool, to identify handles and avoid mix-ups
  • No tools required to attach to the pipette
  • Handle is autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material Polyethylene
Sterility Non-sterile
Graduations None
Quantity 3 Handles
Color White
Length (Metric) 76.2 in
Type Handle
Disposable Reusable
For Use With (Equipment) Gilson PIPETMAN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.