missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ G
GREENER_CHOICE

Product Code. 16343186 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Number of Channels:
8
12
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Number of Channels Unit Size
16343186 2 to 20 μL 12 Each
16313186 1 to 10 μL 8 Each
16323186 1 to 10 μL 12 Each
16333186 2 to 20 μL 8 Each
16353186 20 to 200 μL 8 Each
16363186 20 to 200 μL 12 Each
16373186 30 to 300 μL 8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16343186 Supplier Gilson™ Supplier No. F144071
Powering Proteomics
Use promo code 28003
View all promotions
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PIPETMAN™ G offers a low pipetting force and hours of comfort due to a redesigned piston assembly system.

  • With a redesigned piston assembly system and high quality piston grease, PIPETMAN™ G offers hours of comfortable pipetting
  • PIPETMAN™ G relies on the legendary accuracy, precision, and reliability of PIPETMAN™ with improved pipetting comfort
  • Made of PVDF and stainless steel
  • Offers premium durability
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Declaration of Conformity
Autoclavable Not Autoclavable
Color White
Material Metal Ejector
Model P12x20G
Number of Channels 12
Product Line Pipetman G
Type Manual Air Displacement Pipette
Volume (Metric) 2 to 20 μL
Warranty 3 Years
Length (English) 6.02 in.
Width (English) 12.87 in.
Height (English) 1.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.