missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Pipette Basket

Product Code. 11971501 Shop All Bürkle Products
Change view
Click to view available options
Height (Metric):
280 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
11971501 280 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11971501 Supplier Buerkle™ Supplier No. 96500653
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The Pipette Basket is an accessory for the pipette rinser. For easy and gentle cleaning and safe storage of pipettes.

  • For easy and gentle cleaning and safe storage of pipettes
  • Made of PE
  • Effective height 600 mm
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) For Use with The Pipette Rinser
Product Type Pipet Basket
Height (Metric) 280 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.