missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Pipette Basket

Product Code. 11310455
Change view
Click to view available options
Height (Metric):
280 mm basket, 495 mm total
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
11310455 280 mm basket, 495 mm total Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11310455 Supplier Kartell™ Supplier No. 0022200
  • ONLINE EXCLUSIVE PRICE
    33.85 €
    31.44 € / Pack of 1
    Save 2.41 €
    7% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kartell™ Pipette Basket is for use with a Kartell™ Automatic Pipette Washer and is designed to minimize breakage as it buffers the tips of pipettes.

  • Houses pipettes
  • Specially designed baskets that minimize breakages as it buffers the tips of pipettes against the soft PE base

Specifications

Color White
For Use With (Application) Pipettes and burettes
Quantity 1
Height (Metric) 280 mm basket, 495 mm total
Diameter (Metric) Opening 145 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.