Learn More
Hirschmann™ Pipette Bulbs
Product Code. 11894730
Change view
Click to view available options
Volume (Metric):
0.5 to 100 μL
Unit Size:
Pack of 10
Product Code. Volume (Metric) Unit Size
11894730 0.5 to 100 μL Pack of 10
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11894730 Supplier Hirschmann™ Supplier No. 9650101
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
Type Bulb
Volume (Metric) 0.5 to 100 μL
For Use With (Application) holding and draining disposable micropipettes
Includes rubber cap