Learn More
Sartorius™ Pipette Calibration Kit
Product Code. 14811081 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Cubis Balances
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
14811081 Cubis Balances Each
1 options
Product Code. 14811081 Supplier Sartorius™ Supplier No. YCP04MS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for models with 0.1 mg and 0.01 mg readability.
  • Used for models with 0.1 mg and 0.01 mg readability
  • Consists of moisture trap and all required adapters
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Kit
For Use With (Equipment) Cubis Balances