missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Pipette Cone for Accumax Pipette Controller
Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Green
Orange
Red
Unit Size:
Each
Specifications
Specifications
| Color | Blue |
| Type | Cone |
| For Use With (Application) | Pipetting |
| For Use With (Equipment) | Accumax Pipette Controller |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction