missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Pipette Cone for Accumax Pipette Controller

Product Code. 15870043 Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Green
Orange
Red
Unit Size:
Each

Product Code. 15870043

Only null left
Add to basket
Edge
Add to basket

Available in a range of colors.

  • Autoclavable
  • For use with the Accumax Pipette Controller
  • Pick between blue, red, orange or green

Specifications

Color Red
Type Cone
For Use With (Application) Pipetting
For Use With (Equipment) Accumax Pipette Controller
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.