Learn More
Microspec™ Pipette Tips
Product Code. 16685462
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1000 μL
5 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 300
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility unitSize
16685462 10 μL Non-sterile Pack of 1000
16605472 200 μL Non-sterile Pack of 1000
16615472 1000 μL Non-sterile Pack of 1000
16625472 5 mL Non-sterile Pack of 300
16695462 5 mL Sterile Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16685462 Supplier Microspec™ Supplier No. AE1
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Available as sterile or non-sterile
  • Available as sterile or non-sterile
  • In bulk bags or in grip seal bags
  • Non-filtered
Specifications
Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Blue
Autoclavable Not Autoclavable
Packaging Bulk Bag
Quantity 1000
Disposable Single-use