missing translation for 'onlineSavingsMsg'
Learn More

Pirenoxine, TRC

Product Code. 30138126
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138126 25 mg 25mg
30004681 10 mg 10mg
30010180 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138126 Supplier Toronto Research Chemicals Supplier No. TRCP50858025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1043-21-6
Molecular Formula C16H8N2O5
Molecular Weight (g/mol) 308.25
Synonym 1-Hydroxy-3-carboxy-5H-pyrido[3,2-a]phenoxazin-5-one 1-Hydroxy-5H-pyrido[3,2-a]phenoxazin-5-one-3-carboxylic acid, Catalin, Catalin (pharmaceutical), Catalin K, Clarvisan, HPPC, Kary-Uni, Pirfenoxone, Pyrphenoxone
IUPAC Name 1-hydroxy-5-oxopyrido[3,2-a]phenoxazine-3-carboxylic acid
SMILES O=C1C2=NC(C(O)=O)=CC(O)=C2C3=NC4=C(C=CC=C4)OC3=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H8N2O5/c19-9-5-8(16(21)22)18-14-10(20)6-12-15(13(9)14)17-7-3-1-2-4-11(7)23-12/h1-6H,(H,18,19)(H,21,22)
Formula Weight 308.0433
Chemical Name or Material Pirenoxine
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.