missing translation for 'onlineSavingsMsg'
Learn More

Pirenzepine, Dihydrochloride, TRC

Product Code. 30422284
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422284 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422284 Supplier Toronto Research Chemicals Supplier No. TRCP5085005G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pirenzepine','Dihydrochloride
CAS 29868-97-1
Purity Grade Notes HPLC
Molecular Formula C19 H21 N5 O2 . 2 Cl H
Quantity 5 g
Synonym 6H-Pyrido[2,3-b][1,4]benzodiazepin-6-one','5,11-dihydro-11-[2-(4-methyl-1-piperazinyl)acetyl]-','hydrochloride (1:2), 6H-Pyrido[2,3-b][1,4]benzodiazepin-6-one','5,11-dihydro-11-[(4-methyl-1-piperazinyl)acetyl]-','dihydrochloride (8CI,9CI), 5,11-Dihydro-11-[(4-methyl-1-piperazinyl)acetyl]-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one dihydrochloride, Duogastral, Durapirenz, Gasteril, Gastricur, Gastrozepin, L-S 519, LS 519Cl2, LS 59Cl2, Leblon, Maghen, Pirenzepine dihydrochloride, Pirenzepine hydrochloride, Renzepin, Tabe, Ulcosan, Ulcuforton
InChI Formula InChI=1S/C19H21N5O2.2ClH/c1-22-9-11-23(12-10-22)13-17(25)24-16-7-3-2-5-14(16)19(26)21-15-6-4-8-20-18(15)24;;/h2-8H,9-13H2,1H3,(H,21,26);2*1H
SMILES Cl.Cl.CN1CCN(CC(=O)N2c3ccccc3C(=O)Nc4cccnc24)CC1
IUPAC Name 11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
Molecular Weight (g/mol) 424.32
Formula Weight 423.1229
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.