missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pirfenidone
GREENER_CHOICE

Product Code. 18762794
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18762794 10 mg 10mg
18702784 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18762794 Supplier Tocris Bioscience™ Supplier No. 1093/10
  • ONLINE EXCLUSIVE PRICE
    177.00 €
    166.95 € / 10mg
    Save 10.05 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Antifibrotic agent; regulates cytokine levels in vivo

Antifibrotic agent, effective in models of pulmonary and lung fibrosis. Inhibits collagen production and fibroblast proliferation. Regulates cytokine levels following oral administration in vivo. Potent scavenger of free radicals and inhibitor of lipid peroxidation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53179-13-8
Molecular Formula C12H11NO

Specifications

Quantity 10 mg
Formula Weight 185.23
Purity >99%
Molecular Weight (g/mol) 185.226
Chemical Name or Material Pirfenidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.