missing translation for 'onlineSavingsMsg'
Learn More

Piroctone olamine, Thermo Scientific™

Product Code. 17710639 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17710639 5 g 5g
17700639 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17710639 Supplier Thermo Scientific Acros Supplier No. 468820050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Alpha Vector PIROCTONEOLAMINE
Chemical Name or Material Piroctone olamine
CAS 68890-66-4
Melting Point 130°C to 138°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C14 H23 N O2 . C2 H7 N O
MDL Number 01690792
Quantity 5 g
Merck Index XIII 7585
Molecular Weight (g/mol) 298.46
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.