missing translation for 'onlineSavingsMsg'
Learn More

Piromidic Acid, TRC

Product Code. 30138423
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138423 100 mg 100mg
17961525 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138423 Supplier Toronto Research Chemicals Supplier No. TRCP509500100MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19562-30-2
Molecular Formula C14 H16 N4 O3
Molecular Weight (g/mol) 288.3
Synonym 8-Ethyl-5,8-dihydro-5-oxo-2-(1-pyrrolidinyl)pyrido[2,3-d]pyrimidine-6-carboxylic acid, 5,8-Dihydro-8-ethyl-5-oxo-2-pyrrolidinopyrido[2,3-d]pyrimidine-6-carboxylic acid, 8-Ethyl-5,8-dihydro-5-oxo-2-pyrrolidinopyrido[2,3-d]pyrimidine-6-carboxylic acid, Bactramyl, Enterol, Gastrurol, NSC 291120, PD 93, Panacid, Piromidic acid, Pirudal, Purim, Pyromidic acid, Reelon, Septural, Uropir
IUPAC Name 8-ethyl-5-oxo-2-pyrrolidin-1-ylpyrido[2,3-d]pyrimidine-6-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2cnc(nc12)N3CCCC3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H16N4O3/c1-2-17-8-10(13(20)21)11(19)9-7-15-14(16-12(9)17)18-5-3-4-6-18/h7-8H,2-6H2,1H3,(H,20,21)
Formula Weight 288.1222
Percent Purity >95%
Chemical Name or Material Piromidic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.