missing translation for 'onlineSavingsMsg'
Learn More

Pivaldehyde, 96%

Product Code. 10703803 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10703803 100 g Glass bottle 100g
10354122 5 g Glass bottle 5g
10682233 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10703803 Supplier Thermo Scientific Acros Supplier No. 368361000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 630-19-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-19-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006962
InChI Key FJJYHTVHBVXEEQ-UHFFFAOYSA-N
Synonym trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
PubChem CID 12417
IUPAC Name 2,2-dimethylpropanal
SMILES CC(C)(C)C=O

Specifications

Melting Point 6.0°C
Color Colorless
Density 0.7750g/mL
Boiling Point 75.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCHO
Refractive Index 1.3775 to 1.3795
Quantity 100 g
Beilstein 01, IV, 3295
Fieser 13,249
Merck Index 15, 7624
Specific Gravity 0.775
Solubility Information Solubility in water: negligibly soluble.
Formula Weight 86.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Pivaldehyde, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pivaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.