missing translation for 'onlineSavingsMsg'
Learn More

Pivalic Acid, TRC

Product Code. 30414975
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414975 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414975 Supplier Toronto Research Chemicals Supplier No. TRCP52000010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Dimethylpropionic acid
CAS 75-98-9
Molecular Formula C5 H10 O2
Quantity 10 g
Synonym Propanoic acid','2,2-dimethyl- (9CI','ACI), 2,2-Dimethylpropanoic acid (ACI), Pivalic acid (7CI','8CI), 2,2,2-Trimethylacetic acid, 2,2-Dimethylpropionic acid, Neopentanoic acid, Neovaleric acid, NSC 65449, tert-Pentanoic acid, Trimethylacetic acid, Trimethylmethanecarboxylic acid, Versatic 5 acid, Versatic Acid 5, α,α-Dimethylpropionic acid
InChI Formula InChI=1S/C5H10O2/c1-5(2,3)4(6)7/h1-3H3,(H,6,7)
SMILES CC(C)(C)C(=O)O
IUPAC Name 2,2-dimethylpropanoic acid
Molecular Weight (g/mol) 102.13
Formula Weight 102.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.