missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PIWIL3 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1166
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PIWI-L3 Synthetic Peptide, PEP-1166, from Invitrogen.
This peptide corresponds to 18 amino acids near the amino terminus of human PIWI-L3.PEP-1166 can be used as a blocking peptide with polyclonal antibody PA5-21052.Specifications
440822 | |
purified | |
Human PIWIL3 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q7Z3Z3 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
PIWIL3 | |
0.02% sodium azide | |
50 μg | |
PBS |