missing translation for 'onlineSavingsMsg'
Learn More

Pizotifen N-Oxide, TRC

Product Code. 30132952
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132952 50 mg 50mg
30132569 100 mg 100mg
30005843 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132952 Supplier Toronto Research Chemicals Supplier No. TRCP55280550MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28588-48-9
Molecular Formula C19 H21 N O S
Molecular Weight (g/mol) 311.44
Synonym 4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-piperidine 1-Oxide;
IUPAC Name 4-(4,5-dihydrobenzo[1,2]cyclohepta[3,4-b]thiophen-10-ylidene)-1-methyl-1$l^{5}-azinane 1-oxide
SMILES CN1(=O)CCC(=C2c3ccccc3CCc4sccc24)CC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H21NOS/c1-20(21)11-8-15(9-12-20)19-16-5-3-2-4-14(16)6-7-18-17(19)10-13-22-18/h2-5,10,13H,6-9,11-12H2,1H3/b19-15-
Formula Weight 311.1344
Chemical Name or Material Pizotifen N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.