missing translation for 'onlineSavingsMsg'
Learn More

Pizotyline, TRC

Product Code. 17933705
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933705 500 mg 500mg
30137276 50 mg 50mg
30135183 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933705 Supplier Toronto Research Chemicals Supplier No. TRCP552800500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15574-96-6
Formula Weight 295.1395
InChI Formula InChI=1S/C19H21NS/c1-20-11-8-15(9-12-20)19-16-5-3-2-4-14(16)6-7-18-17(19)10-13-21-18/h2-5,10,13H,6-9,11-12H2,1H3
IUPAC Name 4-(4,5-dihydrobenzo[1,2]cyclohepta[2,4-b]thiophen-10-ylidene)-1-methylpiperidine
Molecular Formula C19 H21 N S
Molecular Weight (g/mol) 295.44
SMILES CN1CCC(=C2c3ccccc3CCc4sccc24)CC1
Synonym Piperidine, 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-, 4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine, 4H-Benzo[4,5]cyclohepta[1,2-b]thiophene, piperidine deriv., 4-(1-Methyl-4-piperidylidene)-9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophene, BC 105, Litec, Pizotifen, Pizotyline, Polomigran, Sandomigran
Chemical Name or Material Pizotyline

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.