missing translation for 'onlineSavingsMsg'
Learn More

pJTI™ R4 Exp CMV EmGFP pA Vector

Product Code. 11681869
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11681869 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11681869 Supplier Invitrogen Supplier No. A14146
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

pJTI™ R4 Exp CMV EmGFP pA Vector

The pJTI™ R4 Exp CMV EmGFP pA vector is a positive control vector for assessing the success of a retargeting reaction with a Jump-In™ parental cell line. When co-transfected with the integrase vector (pJTI™ R4 Int vector) included in the Jump-In™ parental kits, the EmGFP will be expressed and positive cells will fluoresce green.

Ensure the Success of Your Jump-In™ Retargeting Reactions
Successful retargeting of Jump-In™ parental cell lines like the the Jump-In™ GripTite™ HEK293 kit (A14150) is dependent on a variety of factors such as:

Transfection efficiency

  • Cell confluency
  • Antibiotic selection conditions
  • Quality and concentration of DNA
  • Retargeting vector to integrase vector ratio

    We strongly recommend including a retargeting reaction with the pJTI™ R4 Exp CMV EmGFP pA vector in your Jump-In™ experiment along with negative controls (no plasmid DNA, no integrase vector) so you can easily visualize the results and optimize the retargeting conditions.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Constitutive or Inducible System Constitutive
    Delivery Type Transfection
    Format Liquid
    Promoter CMV
    Product Type Mammalian Expression Vector
    Selection Agent (Eukaryotic) None
    Content And Storage Immediately upon receipt, store vector at -20°C
    Shipping Condition Dry Ice
    Protein Tag Untagged
    Cloning Method Gateway
    Quantity 100 μg
    Vector Jump-In Vectors
    Product Line Jump-In
    For Use With (Application) Stable Cell Line Development, Targeted Integration
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.