missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pJTI™ R4 EXP CMV-TO EmGFP pA Vector
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
The pJTI™ R4 Exp CMV-TO EmGFP pA vector is a positive control vector for assessing retargeting efficiency when retargeting a Jump-In™ T-REx™ parental cell line. When co-transfected with the integrase vector (pJTI™ R4 Int vector included in the Jump-In™ parental kits) and after antibiotic selection, the EmGFP can be inducibly expressed with doxycycline and the successfully retargeted cells will fluoresce green.
Ensure the Success of Your Jump-In™ T-REx™ Retargeting Reactions
Successful retargeting of Jump-In™ T-REx™ parental cell lines like the Jump-In™ T-REx™ HEK293 Kit is dependent on a variety of factors, such as:
Transfection efficiency
Cell confluency
Antibiotic selection conditions
Quality and concentration of DNA
Retargeting vector to integrase vector ratio
We strongly recommend including a positive control retargeting reaction using the pJTI™ R4 Exp CMV-TO EmGFP pA vector in your Jump-In™ experiment, along with negative controls (no plasmid DNA, no integrase vector), so you can easily visualize the results and optimize the retargeting conditions.
Ensure the Success of Your Jump-In™ T-REx™ Retargeting Reactions
Successful retargeting of Jump-In™ T-REx™ parental cell lines like the Jump-In™ T-REx™ HEK293 Kit is dependent on a variety of factors, such as:
We strongly recommend including a positive control retargeting reaction using the pJTI™ R4 Exp CMV-TO EmGFP pA vector in your Jump-In™ experiment, along with negative controls (no plasmid DNA, no integrase vector), so you can easily visualize the results and optimize the retargeting conditions.
Specifications
Specifications
| Constitutive or Inducible System | Inducible |
| Delivery Type | Transfection |
| Inducing Agent | Tetracycline |
| Promoter | CMV/TO |
| Product Type | Mammalian Expression Vector |
| Reporter Gene | GFP (EmGFP) |
| Selection Agent (Eukaryotic) | Geneticin™ (G-418) |
| Content And Storage | pJTI™ R4 EXP CMV-TO EmGFP pA vector (100 μg), store at -20 °C |
| Antibiotic Resistance Bacterial | Ampicillin (AmpR) |
| Protein Tag | GFP (EmGFP) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction