missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PKUMDL WQ 2101

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
PKUMDL WQ 2101 is a negative allosteric modulator of phosphoglycerate dehydrogenase (PHGDH) (IC50 = 34.8 μM). Exhibits antitumor activity in vitro in breast cancer cells lines overexpressing PHGDH (EC50 values are 7.7 and 10.8 μM in MDA-MB-468 and HCC-70 cells, respectively). Inhibits growth of MDA-MB-468 xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydroxy-N'-(2-hydroxy-5-nitrobenzylidene)benzohydrazide |
| CAS | 304481-72-9 |
| Quantity | 5 mg |
| Molecular Formula | C14H11N3O6 |
| Formula Weight | 317.25 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction