missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PKUMDL WQ 2101

Description
PKUMDL WQ 2101 is a negative allosteric modulator of phosphoglycerate dehydrogenase (PHGDH) (IC50 = 34.8 μM). Exhibits antitumor activity in vitro in breast cancer cells lines overexpressing PHGDH (EC50 values are 7.7 and 10.8 μM in MDA-MB-468 and HCC-70 cells, respectively). Inhibits growth of MDA-MB-468 xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydroxy-N'-(2-hydroxy-5-nitrobenzylidene)benzohydrazide |
| CAS | 304481-72-9 |
| Quantity | 25 mg |
| Molecular Formula | C14H11N3O6 |
| Formula Weight | 317.25 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?