missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVOS™ Light Cube, Cy5.5 2.0
Specifications
Specifications
| Type | LED Light Cube |
| Product Line | EVOS |
| Excitation/Emission | Ex: 655/40 nm Em: 725/40 |
| For Use With (Equipment) | Countess™ III FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System |
| Includes | Condensing Optics, Hard-coated Filters |
| Quantity | 1 each |
Frequently Asked Questions (FAQs)
Are EVOS 2.0 Light Cubes compatible with the Countess II FL?
Yes. All our EVOS Light Cubes can be used with the Countess II FL.
Are EVOS 2.0 Light Cubes compatible with older EVOS instruments?
Yes. Our EVOS 2.0 Light cubes can be used in all EVOS instruments with EVOS light cube functionality.
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?