missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plant RNA Lysis Solution

Product Code. 15612032 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15612032 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15612032 Supplier Thermo Scientific™ Supplier No. R1641
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific GeneJET Plant RNA Lysis Solution is a component of the GeneJET Plant RNA Purification Mini Kit and may be purchased separately. Provided in 200 mL aliquots.

Thermo Scientific GeneJET Plant RNA Lysis Solution is a component of the GeneJET Plant RNA Purification Mini Kit (Cat. No. K0801/K0802) and may be purchased separately. Provided in 200 mL aliquots.
TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Product Line GeneJET
Quantity 200 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.