missing translation for 'onlineSavingsMsg'
Learn More

Abcam Plasma Fluorescent Exosome Standard, Liquid, 100 μL

Product Code. 32108803
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32108803 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32108803 Supplier Abcam Supplier No. ab288123100uL
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Standardized plasma exosomes for EV tracking in fluorescence microscopy experiments.

  • Applications: Fluorescence Microscopy, Flow Cyt, EM
  • Form: Liquid

Specifications

Alternate Part Number M1073
Content And Storage Store at -20°C.
Form Liquid
Shipping Condition Blue Ice
Target Molecule Exosomes
Detection Method Fluorescence
Sample Type Plasma
Quantity 100 μL
For Use With (Application) Fluorescence microscopy, flow cytometry, electron microscopy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.