missing translation for 'onlineSavingsMsg'
Learn More

THOMSON INSTRUMENT COMPANY Plasmid+™ Media, Sterile, 1 L

Product Code. 12644447
Encompass_Preferred
Change view
Click to view available options
Quantity:
6/Cs.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12644447 6/Cs. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12644447 Supplier THOMSON INSTRUMENT COMPANY Supplier No. 446300
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimized microbial growth media that supports high-copy plasmid production while improving culture performance and downstream processing efficiency.

  • Formulation for E. coli growth and high-copy plasmids
  • Supports low endotoxin output to reduce downstream cleanup
  • Consistent, reproducible results across culture runs
  • Works on standard orbital shakers from bench to pilot volumes
  • Made in the USA
  • The Ultra Yield™ System combines Ultra Yield™ Flasks, AirOtop™ Enhanced Seals/Vented Caps, and Plasmid+™ Enriched Media to support microbial productivity-especially E. coli-for plasmid DNA and protein expression. Across customer studies, the system has achieved 85+ supercoiled plasmid DNA with very low endotoxin, delivered 4 to 10X material cost savings with about 50% less FTE time, and shown roughly 10X higher aeration with scalability from 125 mL to 2.5 L. Bidirectional Transfer Caps on 1.5 L/2.5 L flasks support sterile harvests or transfers to larger fermenters.

Specifications

Classification Animal Origin-free
Form Liquid
Product Type Media
Serum Level Serum-free
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 6/Cs.
Culture Type Suspension Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.