missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Plastic Base and Safety Ring

Product Code. p-7162719
Change view
Click to view available options
For Use With (Equipment):
Measuring Cylinder
Measuring Cylinders Without Base (1000 mL)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11776714 Measuring Cylinder Each
10099790 Measuring Cylinders Without Base (1000 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11776714 Supplier Hirschmann™ Supplier No. 8900160
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Plastic socket for safe standing and protection of graduated cylinders.

  • For measuring cylinders
  • Without base
  • Available to fit a range of sizes

Specifications

Color Green
Capacity (Metric) 10 mL
For Use With (Equipment) Measuring Cylinder
No. per Pack 1 Each
Includes Protecting Ring
Type Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.