missing translation for 'onlineSavingsMsg'
Learn More

Manutan Plastic chains, 25m, yellow/black plastic chain, 10mm diameter

Product Code. 12871487
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12871487 Supplier Manutan Supplier No. A011636
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High density polyethylene chain. UV stabilised, can easily be cut to required length.

  • Length : 25m
  • Yellow/Black
  • Diameter : 10mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.