Learn More
Fisherbrand™ Plastic Covers with Orifice for S 50 R Ultrasonic Cleaning Units
Product Code. 12699345 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Bath, S 50 R
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12699345 Ultrasonic Bath, S 50 R Each
1 options
Product Code. 12699345
  • ONLINE EXCLUSIVE PRICE
    31.85 €
    25.72 € / Pack of 1
    Save 6.13 €
    19% Off
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Allows glass beakers with a diameter between 90 and 97mm to be inserted into the bath for direct ultrasonic treatment. Fisherbrand™ Plastic Covers with Orifice for S 50 R Ultrasonic Cleaning Units can be combined with a rubber ring to securely and safely hold the beaker during the application.
  • Manufactured from durable blue plastic
Specifications
Type Lid
For Use With (Equipment) Ultrasonic Bath, S 50 R
Material Plastic