Learn More
Microspec™ Plastic Culture Loop
Product Code. 17845718
Change view
Click to view available options
Volume:
1 μL
5 μL
10 μL
Unit Size:
Pack of 12000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume unitSize
17845718 1 μL Pack of 12000
17855718 5 μL Pack of 12000
17835718 10 μL Pack of 12000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17845718 Supplier Microspec™ Supplier No. C2G
Only null left
Add to basket
Add to basket
Microseal pouches of sterile plastic culture loops.
  • 12,000 Plastic Culture Loops
  • Microseal Pouch of 20
  • 200 mm length
  • Sterile EtO
Specifications
Disposable Single-use
Volume 1 μL
Length (English) 7.8 in.
Length (Metric) 200 mm
Material Plastic
Sterility Sterile
Size Standard
Color Blue
Product Type Inoculating Loop
Calibrated Yes
Autoclavable Not Autoclavable