missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Plastic Culture Needles

Product Code. 15782125
Change view
Click to view available options
Sterility:
Aseptically Manufactured
Gamma-irradiated
Quantity:
1000
16000
Unit Size:
Pack of 1000
Pack of 16000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Quantity unitSize
15782125 Aseptically Manufactured 1000 Pack of 1000
15762125 Aseptically Manufactured 16000 Pack of 16000
15792125 Gamma-irradiated 1000 Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15782125 Supplier Microspec™ Supplier No. F4
  • ONLINE EXCLUSIVE PRICE
    45.21 €
    39.14 € / Pack of 1000
    Save 6.07 €
    13% Off
In Stock
Add to basket
Edge
Add to basket

Transfer and inoculate living microorganisms

  • Sharp end to facilitate easy stab inoculation
  • Picks up small colonies
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured

Specifications

Disposable Single-use
Length (English) 7.9 in.
Length (Metric) 200 mm
Material High-impact Polystyrene
Sterility Aseptically Manufactured
Color Blue
Product Type Inoculating Needle
Quantity 1000
Packaging Type Packaged in Microseal Pouches of 20
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.