missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Plastic, Jerry Can

Product Code. 10083084
Change view
Click to view available options
Capacity (Metric):
25 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10083084 25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083084 Supplier Kautex™ Supplier No. 2000086793
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Color: Natural

  • Plastic
  • Containers
  • without caps
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 6.6 gal.
Capacity (Metric) 25 L
Color Natural
Material Plastic
Product Type Jerry Can
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.