Learn More
Pacific Handy Cutter Plastic Spring Back Safety Knife
Product Code. 17738977
Change view
Click to view available options
Color:
Yellow
Unit Size:
Pack of 50
Product Code. Color Unit Size
17738977 Yellow Pack of 50
1 options
Product Code. 17738977 Supplier Pacific Handy Cutter Supplier No. EZ3
  • ONLINE EXCLUSIVE PRICE
    401.51 €
    346.46 € / Pack of 50
    Save 55.05 €
    13% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The EZ-3 is a lightweight safety knife with automatic blade retraction. Thus, it offers a high level of security.
  • Due to the plastic body, it is particularly economical.
  • Changing the blade is very easy, and the knife handle can be opened completely for cleaning.
  • Additional blades can be stowed away in the knife handle.
  • Short blade length, therefore suitable for light cutting activities.
  • Knife in clearly visible yellow
Specifica
Product Type Spring Back Safety Knife
Disposable Reusable
Sterility Non-sterile
Color Yellow
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.