missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Micronova Plastic Squeegees easily clean smooth or uneven surfaces and minimize contamination areas by reducing dirt and bacteria growth. Designed for use with STAH or STAEH series plastic handles. Available in a variety of colors.
- Durably constructed polypropylene squeegees
- Choice of solid rubber or double foam blades
- Excellent for use on smooth surfaces and uneven floor surfaces
- Prevents hazardous sparks when cleaning-up flammable fluids
- Available in 18 and 24 inch (45 and 60 cm) widths
- Use with STAH or STAEH handles for a secure fit
- Will not rust
Specifications
Specifications
| Product Type | Squeegee, ACME Threaded End |
| Color | Dark Blue |
| Content And Storage | Store in cool dry area when not in use. |
| Dimensions (L x W) | 61 x 5.1 cm (24 x 2 in.) |
| Disposable | Reusable |
| For Use With (Application) | Squeegee for Floors, Walls and Windows |
| For Use With (Equipment) | STA Series Plastic Handles |
| Material | Rubber Blade, Polypropylene Frame |
| Width (English) | 2 in. |
| Width (Metric) | 5.1 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?