Learn More
Kartell™ Plastilab™ Tapered Straight Connectors
Product Code. 11924349
Click to view available options
Diameter (Metric):
4-6 mm
6-8 mm
8-10 mm
10-12 mm
12-14 mm
Packaging Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 11924349
Brand: Kartell™ 7051000
Only null left
Add to basket
Edge
Add to basket
For straight connection of tubing with different diameters.
For straight connection of tubings with different diameters.
  • Constructed from polypropylene
  • Autoclavable
Specifications
Diameter (Metric) 4-6 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) For straight connection of different diameter tubing
Packaging Quantity 10/Pk.
Port Size 2mm bore
Shape Straight
Product Type Straight Tapered Tubing Connector