Learn More
Kartell™ Plastilab™ Untapered T-Shaped Tubing Connectors
BIOPHARMA
Designed with serrations to hold hoses tightly and prevent slipping.
Brand:  Kartell™ 7046000
Product Code. 11994349
  • ONLINE EXCLUSIVE PRICE
    6.00 €
    4.90 € / Pack of 10
    Save 1.10 €
    18% Off
In Stock
Edge
Add to basket
Description
Description
Designed with serrations to hold hoses tightly and prevent slipping.
  • Made from polypropylene
  • Autoclavable
  • Chemical- and temperature-resistant
Compliance
Meet Food and Agriculture regulations