Learn More
Eppendorf™ Plate Carriers for Buckets
Product Code. 15748379 Shop All Eppendorf Products
Change view
Click to view available options
Product Type:
Plate Carrier for High-capacity Bucket
Plate Carrier for Plate/Tube Bucket
Capacity:
6/24 (MTP)\n2/8 (DWP)
7/28 (MTP)\n2/8 (DWP)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Capacity unitSize
15748379 Plate Carrier for Plate/Tube Bucket 7/28 (MTP)\n2/8 (DWP) Each
15684614 Plate Carrier for High-capacity Bucket 6/24 (MTP)\n2/8 (DWP) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15748379 Supplier Eppendorf™ Supplier No. 5920705008
In Stock
Add to basket
Add to basket
Carriers designed for MTP, DWP, PCR plates. Available for high-capacity buckets and plate/tube buckets
  • Number of tubes per adapter/per rotor: 7/28(MTP)\n2/8(DWP) or 6/24 (MTP)\n2/8 (DWP)
  • Set of 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) for MTP, DWP, PCR plates
Capacity 7/28 (MTP)\n2/8 (DWP)
Product Type Plate Carrier for Plate/Tube Bucket