Learn More
Kinematica™ Plate Stand ST-P15/320 for Polytron™ 1300D Homogenizer
Product Code. 13467639 Shop All Kinematica Products
Change view
Click to view available options
For Use With (Equipment):
PT 1300 D Disperser
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
13467639 PT 1300 D Disperser Each
1 options
Product Code. 13467639 Supplier Kinematica™ Supplier No. 11040013
  • ONLINE EXCLUSIVE PRICE
    409.50 €
    378.00 € / Pack of 1
    Save 31.50 €
    7% Off
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stainless steel stand with base, height-adjustable bracket for supporting the drive and base mounting.
With height-adjustable holder for drive support stainless steel for use with Polytron™ PT 1300 D drive.
Specifications
Product Type Plate Stand
Description Stainless steel stand with base, height-adjustable bracket for supporting the drive and base mounting
For Use With (Equipment) PT 1300 D Disperser
Includes The safety ring prevents the uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height.
Model ST-P 15/320