missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ Platform for Grant PSU10i Oribtial Shaker
Includes clamps to keep flasks and beakers from spilling. Grant Instruments™ Platform for Grant Oribtial Shakers and Shaker Incubators are interchangeable to accommodate a variety of vessels.
265.00 € - 283.02 €
Specifications
Dimensions (L x W) | 250 x 190 mm |
---|---|
For Use With (Equipment) | PSU-10i Orbital Platform Shaker |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11461058
|
Grant Instruments™
P6-250 |
6 x 250 to 300 mL flasks and beakers |
265.00 €
Each |
In Stock
Log in to see stock. |
|||||
11451058
|
Grant Instruments™
P12-100 |
12 x 100 to 150 mL flasks and beakers |
283.02 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Includes clamps to keep vessels secure
- For use with
- ES-20 Shaker Incubator
- ES-80 Shaker Incubator
- PSU 10i Orbital Shaker
- PSU 20i Orbital Shaker
Specifications
250 x 190 mm | |
PSU-10i Orbital Platform Shaker |