missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Platform trolley, steel, Ekwo, 300kg capacity

Brand:  Key Industrial Equipment™ A042465

Encompass_Preferred

Product Code. 12827548

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Durable steel trolley with a push/pull handle that folds for easy storage and carriage. All round PVC buffer strip protects wall and door frames. Non-slip PVC platform surface. Ideal for the warehouse or office.
  • Dimensions (W x L): 610mm x 910mm
  • Fitted with two fixed and two rear swivel castors for easy manoeuvring.
  • Handle height: 870mm
  • Load capacity: 300kg
  • Weight: 16.7kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.