missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 0.5% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced

Product Code. 11399379
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399379 100 g 100g
11309389 25 g 25g
11389379 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399379 Supplier Thermo Scientific Alfa Aesar Supplier No. 089106.22
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-6-6

Platinum, 0.5% on 2.7-3.3mm (0.11-0.13in) alumina pellets is used for selective hydrogenations. It is also used in gas purifications, especially oxidation of carbon monoxide to carbon dioxide and hydrogen removal from oxygen or carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective hydrogenations; gas purifications: e
g
  • Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
  • TRUSTED_SUSTAINABILITY

    Chemical Identifiers

    CAS 6-6-7440
    MDL Number MFCD00011179

    Specifications

    Quantity 100 g
    Solubility Information Insoluble in water.
    Chemical Name or Material Platinum, 0.5% on alumina pellets, 2.7 to 3.3mm (0.11 to 0.13 in.), Reduced

    RUO – Research Use Only

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.