missing translation for 'onlineSavingsMsg'
Learn More

Platinum Cobalt Colour 10 Color Standard, Lovibond™

Catalog Number 11987785
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Quantity 500 mL
Odor Odorless
Solution Contains Hexachloroplatinate of Potassium, Cobalt chloride, Hydrochloric acid
pH ∼3
Solubility Miscible with water
Physical Form Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'