missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Cobalt Colour 30 Color Standard, Lovibond™
Click to view available options
Quantity:
500 mL
Specifications
Quantity | 500 mL |
Odor | Odorless |
Solution Contains | Hexachloroplatinate of Potassium, Cobalt chloride, Hydrochloric acid |
pH | ∼3 |
Solubility | Miscible with water |
Physical Form | Solution |
Safety and Handling
Product Identifier
- 500ML Platinum-Cobalt Colour 30
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'